accessibility menu, dialog, popup

UserName

CAS RN 16639-91-1

H3COOHNONH2OHO

CAS RN 16639-91-1

IUPAC Name: 3-carbamoyl-2-[(ethoxycarbonyl)amino]propanoic acid
Synonyms: 3-carbamoyl-2-[(ethoxycarbonyl)amino]propanoic acid
Molecular Weight (g/mol): 204.18
Molecular Formula: C7H12N2O5
InChi Key: YORYXPTWPIKBDE-UHFFFAOYNA-N
SMILES: CCOC(=O)NC(CC(N)=O)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)