CAS RN 1660963-42-7
CAS RN 1660963-42-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Olafertinib, MedChemExpress
MedChemExpress Olafertinib is a third-generation EGFR TKI, with GI50 values of 5 nM (EGFR L858R/T790M), 10 nM (EGFR del19) and 689 nM (EGFR WT), respectively. Olafertinib has the potential for NSCLC research.