Learn More
Olafertinib, MedChemExpress
Product Code. 16493696
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16493696 25 mg 25mg
16470388 5 mg 5mg
16467726 100 mg 100mg
16427928 10 mg 10mg
16423867 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 16493696 Supplier Medchem Express Supplier No. HY19815
MedChemExpress Olafertinib is a third-generation EGFR TKI, with GI50 values of 5 nM (EGFR L858R/T790M), 10 nM (EGFR del19) and 689 nM (EGFR WT), respectively. Olafertinib has the potential for NSCLC research.
Olafertinib is a third-generation EGFR TKI, with GI50 values of 5 nM (EGFR L858R/T790M), 10 nM (EGFR del19) and 689 nM (EGFR WT), respectively. Olafertinib has the potential for NSCLC research.
Specifications
Chemical Name or Material Olafertinib
CAS 1660963-42-7
Color Yellow
Purity Grade Notes Research
Molecular Formula C29H28F2N6O2
Quantity 25 mg
Solubility Information DMSO : 125 mg/mL (235.60 mM; Need ultrasonic)
SMILES OCCN1CCN(C2=C(F)C(F)=C(NC3=NC=C4C(C(C5=CC(NC(C=C)=O)=CC=C5)=CC=C4)=N3)C=C2)CC1
Molecular Weight (g/mol) 530.57
Formula Weight 530.57
Percent Purity 99.41%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease