accessibility menu, dialog, popup

UserName

CAS RN 162401-59-4

IUPAC Name: 4-(difluoromethoxy)-3-methoxybenzoic acid
Synonyms: 4-(difluoromethoxy)-3-methoxybenzoic acid
Molecular Weight (g/mol): 218.16
Molecular Formula: C9H8F2O4
InChi Key: QEZBOPWZULAPBJ-UHFFFAOYSA-N
SMILES: COC1=C(OC(F)F)C=CC(=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results