CAS RN 159857-80-4
CAS RN 159857-80-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MedChemExpress MC-Val-Cit-PAB
MedChemExpress MC-Val-Cit-PAB is a cathepsin cleavable ADC linker that is used for making antibody-drug conjugate.