Learn More
MedChemExpress MC-Val-Cit-PAB
Specifications
Specifications
| Purity | 96.57% |
| CAS | 159857-80-4 |
| Content And Storage | 4°C, protect from light, stored under nitrogen∣In solvent : -80°C, 6 months∣-20°C, 1 month (protect from light, stored under nitrogen) |
| For Use With (Application) | Cancer-programmed cell death |
| Molecular Weight (g/mol) | 572.65 g/mol |
| Product Type | Linker |
| Quantity | 1 g |
| Shipping Condition | Room temperature |
| Solubility | DMSO : 190 mg/mL (331.79 mM∣Need ultrasonic) |