CAS RN 157026-18-1
CAS RN 157026-18-1
IUPAC Name:
4-bromo-2-nitro-6-(trifluoromethyl)aniline
Synonyms:
4-bromo-2-nitro-6-(trifluoromethyl)aniline
Molecular Weight (g/mol):
285.02
Molecular Formula:
C7H4BrF3N2O2
InChi Key:
JWIAFKYOENFPNQ-UHFFFAOYSA-N
SMILES:
NC1=C(C=C(Br)C=C1C(F)(F)F)[N+]([O-])=O
1
–
1
of
1
results
1
4-Bromo-2-nitro-6-(trifluoromethyl)aniline, 98%, Thermo Scientific™
CAS: 157026-18-1 Molecular Formula: C7H4BrF3N2O2 Molecular Weight (g/mol): 285.02 MDL Number: MFCD09835318 InChI Key: JWIAFKYOENFPNQ-UHFFFAOYSA-N Synonym: 4-bromo-2-nitro-6-trifluoromethyl aniline,2-amino-5-bromo-3-nitrobenzotrifluoride,4-bromo-2-nitro-6-trifluoromethyl-phenylamine,4-bromo-2-nitro-6-trifluoromethyl benzenamine,4-bromo-2-trifluoromethyl-6-nitrobenzenamine,acmc-209df8,ksc910q9b,benzenamine,4-bromo-2-nitro-6-trifluoromethyl,4-bromo-2-nitro-6-trifluoromethyl-benzenamine PubChem CID: 10850672 IUPAC Name: 4-bromo-2-nitro-6-(trifluoromethyl)aniline SMILES: NC1=C(C=C(Br)C=C1C(F)(F)F)[N+]([O-])=O