Learn More
4-Bromo-2-nitro-6-(trifluoromethyl)aniline, TRC
Specifications
Specifications
| Chemical Name or Material | 4-Bromo-2-nitro-6-(trifluoromethyl)aniline |
| CAS | 157026-18-1 |
| Molecular Formula | C7H4BrF3N2O2 |
| Quantity | 2.5 g |
| Synonym | 4-Bromo-2-nitro-6-(trifluoromethyl)benzenamine, 2-Amino-5-bromo-3-nitrobenzotrifluoride |
| InChI Formula | InChI=1S/C7H4BrF3N2O2/c8-3-1-4(7(9,10)11)6(12)5(2-3)13(14)15/h1-2H,12H2 |
| SMILES | C1=C(C=C(C(=C1C(F)(F)F)N)[N+](=O)[O-])Br |
| IUPAC Name | 4-bromo-2-nitro-6-(trifluoromethyl)aniline |
| Molecular Weight (g/mol) | 285.02 |
| Formula Weight | 283.9408 |