accessibility menu, dialog, popup

UserName

CAS RN 152586-90-8

IUPAC Name: N-(2,4-dimethoxyphenyl)-4-nitrobenzamide
Synonyms: N-(2,4-dimethoxyphenyl)-4-nitrobenzamide
Molecular Weight (g/mol): 302.29
Molecular Formula: C15H14N2O5
InChi Key: XUJKRQHILJWOBB-UHFFFAOYSA-N
SMILES: COC1=CC(OC)=C(NC(=O)C2=CC=C(C=C2)[N+]([O-])=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results