accessibility menu, dialog, popup

UserName

CH3OOSH3COOCH3HNOCl

CAS RN 146381-87-5

IUPAC Name: 2,4-diethyl 5-(2-chloroacetamido)-3-methylthiophene-2,4-dicarboxylate
Synonyms: 2,4-diethyl 5-(2-chloroacetamido)-3-methylthiophene-2,4-dicarboxylate
Molecular Weight (g/mol): 333.78
Molecular Formula: C13H16ClNO5S
InChi Key: AOFYGAIXNQKIHT-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=C(C)C(C(=O)OCC)=C(NC(=O)CCl)S1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results