accessibility menu, dialog, popup

UserName

BOOH3CCH3H3CH3COH

CAS RN 1449133-12-3

IUPAC Name: 3-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]prop-2-yn-1-ol
Synonyms: 3-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]prop-2-yn-1-ol
Molecular Weight (g/mol): 258.12
Molecular Formula: C15H19BO3
InChi Key: SYRFTFKBNBRKRB-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC=C(C=C1)C#CCO

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results