CAS RN 1421261-37-1
IUPAC Name:
methyl 4-(3-ethyl-1,2,4-oxadiazol-5-yl)benzoate
Synonyms:
methyl 4-(3-ethyl-1,2,4-oxadiazol-5-yl)benzoate
Molecular Weight (g/mol):
232.24
Molecular Formula:
C12H12N2O3
InChi Key:
ZEDYJJCOGRSZFM-UHFFFAOYSA-N
SMILES:
CCC1=NOC(=N1)C1=CC=C(C=C1)C(=O)OC
1
–
1
of
1
results
1
Methyl 4-(3-ethyl-1,2,4-oxadiazol-5-yl)benzoate, 97%, Thermo Scientific™
CAS: 1421261-37-1 Molecular Formula: C12H12N2O3 Molecular Weight (g/mol): 232.239 MDL Number: MFCD18384922 InChI Key: ZEDYJJCOGRSZFM-UHFFFAOYSA-N Synonym: methyl 4-3-ethyl-1,2,4-oxadiazol-5-yl benzoate,4-3-ethyl-1,2,4-oxadiazol-5-yl benzic acid methyl ester PubChem CID: 73995175 IUPAC Name: methyl 4-(3-ethyl-1,2,4-oxadiazol-5-yl)benzoate SMILES: CCC1=NOC(=N1)C2=CC=C(C=C2)C(=O)OC