CAS RN 141977-79-9
CAS RN 141977-79-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Miriplatin, MedChemExpress
MedChemExpress Miriplatin (SM-11355) is a chemotherapy agent which belongs to the class of alkylating agents.