Learn More
Miriplatin, MedChemExpress
Product Code. 16487086
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16487086
Brand: Medchem Express HY16325A
MedChemExpress Miriplatin (SM-11355) is a chemotherapy agent which belongs to the class of alkylating agents.
Miriplatin (SM-11355) is a chemotherapy agent which belongs to the class of alkylating agents.
Specifications
Chemical Name or Material Miriplatin
Physical Form Solid
CAS 141977-79-9
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C34H68N2O4Pt
For Use With (Application) Cancer-programmed cell death
Synonym SM-11355
Solubility Information Acetone : < 1 mg/mL (ultrasonic) (insoluble; DMSO can inactivate Miriplatin's activity) ∣DMF : < 1 mg/mL (insoluble; DMSO can inactivate Miriplatin's activity) ∣H2O : < 0.1 mg/mL (insoluble; DMSO can inactivate Miriplatin's activity)
SMILES NC1CCCCC1N.[O-]C(CCCCCCCCCCCCC)=O.[O-]C(CCCCCCCCCCCCC)=O.[Pt+2]
Molecular Weight (g/mol) 764.0
Formula Weight 764.0
Percent Purity 98.0%
Grade Research
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.