CAS RN 140369-67-1
CAS RN 140369-67-1
IUPAC Name:
{4'-fluoro-[1,1'-biphenyl]-4-yl}boronic acid
Synonyms:
4'-fluoro-[1,1'-biphenyl]-4-ylboronic acid
Molecular Weight (g/mol):
216.02
Molecular Formula:
C12H10BFO2
InChi Key:
KHMFYFVXTICBEL-UHFFFAOYSA-N
SMILES:
OB(O)C1=CC=C(C=C1)C1=CC=C(F)C=C1
1
–
1
of
1
results
1
4'-Fluorobiphenyl-4-boronic acid, 98%, Thermo Scientific™
CAS: 140369-67-1 Molecular Formula: C12H10BFO2 Molecular Weight (g/mol): 216.02 MDL Number: MFCD08544387 InChI Key: KHMFYFVXTICBEL-UHFFFAOYSA-N Synonym: 4-4-fluorophenyl phenylboronic acid,4'-fluoro-1,1'-biphenyl-4-yl boronic acid,4'-fluoro-4-biphenylboronic acid,4'-fluorobiphenyl-4-ylboronic acid,4-4-fluorophenyl phenylboronicacid,4'-fluorobiphenyl-4-boronic acid,boronic acid,b-4'-fluoro 1,1'-biphenyl-4-yl,4'-fluoro-1,1'-biphenyl-4-ylboronic acid,akos brn-1038,pubchem11575 PubChem CID: 14985212 IUPAC Name: [4-(4-fluorophenyl)phenyl]boronic acid SMILES: OB(O)C1=CC=C(C=C1)C1=CC=C(F)C=C1