CAS RN 1400591-39-0
IUPAC Name:
4-methylbenzene-1-sulfonic acid; 7-(3-aminopyrrolidin-1-yl)-1-(2,4-difluorophenyl)-6-fluoro-4-oxo-1,4-dihydro-1,8-naphthyridine-3-carboxylic acid
Synonyms:
para-toluene sulfonate; tosufloxacin
Molecular Weight (g/mol):
576.55
Molecular Formula:
C26H23F3N4O6S
InChi Key:
CSSQVOKOWWIUCX-UHFFFAOYNA-N
SMILES:
CC1=CC=C(C=C1)S(O)(=O)=O.NC1CCN(C1)C1=C(F)C=C2C(=O)C(=CN(C3=C(F)C=C(F)C=C3)C2=N1)C(O)=O
1
–
1
of
1
results
1
Tosufloxacin tosylate hydrate, MedChemExpress
MedChemExpress Tosufloxacin (tosylate hydrate) is a fluoroquinolone antibacterial agent.