Learn More
Tosufloxacin tosylate hydrate, MedChemExpress
Product Code. 16498718
Encompass_Preferred
Change view
Click to view available options
Quantity:
200 mg
1 g
5 g
10 g
Unit Size:
10g
1g
200mg
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16498718 10 g 10g
16491128 5 g 5g
16432359 1 g 1g
16411466 200 mg 200mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16498718 Supplier Medchem Express Supplier No. HYB1802A
MedChemExpress Tosufloxacin (tosylate hydrate) is a fluoroquinolone antibacterial agent.
Tosufloxacin (tosylate hydrate) is a fluoroquinolone antibacterial agent.
Specifications
Chemical Name or Material Tosufloxacin tosylate hydrate
Physical Form Solid
CAS 1400591-39-0
Color White
Quantity 10 g
Purity Grade Notes Research
Molecular Formula C26H25F3N4O7S
For Use With (Application) COVID-19-immunoregulation
Synonym A-61827 tosylate hydrate
Solubility Information DMSO : 12.5 mg/mL (21.02 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES CC1=CC=C(S(=O)(O)=O)C=C1.O=C(C2=CN(C3=CC=C(F)C=C3F)C4=C(C=C(F)C(N5CC(N)CC5)=N4)C2=O)O.O
Molecular Weight (g/mol) 594.56
Formula Weight 594.56
Percent Purity 99.03%
Grade Research