accessibility menu, dialog, popup

UserName

CAS RN 1373920-68-3

H3COBrBrOFFF

CAS RN 1373920-68-3

IUPAC Name: 1,5-dibromo-2-methoxy-4-(trifluoromethoxy)benzene
Synonyms: 1,5-dibromo-2-methoxy-4-(trifluoromethoxy)benzene
Molecular Weight (g/mol): 349.93
Molecular Formula: C8H5Br2F3O2
InChi Key: MNJSXTGIUDEJSM-UHFFFAOYSA-N
SMILES: COC1=CC(OC(F)(F)F)=C(Br)C=C1Br

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results