CAS RN 13616-82-5
CAS RN 13616-82-5
IUPAC Name:
2,4-dimethylbenzene-1-thiol
Synonyms:
2,4-dimethylbenzenethiol
Molecular Weight (g/mol):
138.23
Molecular Formula:
C8H10S
InChi Key:
AMNLXDDJGGTIPL-UHFFFAOYSA-N
SMILES:
CC1=CC=C(S)C(C)=C1
1
–
1
of
1
results
1
2,4-Dimethylthiophenol, 95%
CAS: 13616-82-5 Molecular Formula: C8H10S Molecular Weight (g/mol): 138.228 MDL Number: MFCD00010019 InChI Key: AMNLXDDJGGTIPL-UHFFFAOYSA-N Synonym: 2,4-dimethylthiophenol,2,4-dimethyl thiophenol,benzenethiol, 2,4-dimethyl,2,4-thioxylenol,2,4-xylenethiol,4-mercapto-m-xylene,2,4-dimethylbenzene-1-thiol,4-methylthiocresol,4-thio-m-xylene,m-xylene-4-thiol PubChem CID: 83617 IUPAC Name: 2,4-dimethylbenzenethiol SMILES: CC1=CC(=C(C=C1)S)C