CAS RN 1350653-20-1
CAS RN 1350653-20-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Vericiguat, MedChemExpress
MedChemExpress Vericiguat (BAY1021189) is a potent, orally available and soluble guanylate cyclase stimulator.