missing translation for 'onlineSavingsMsg'
Learn More

Vericiguat, MedChemExpress

Product Code. p-200096780
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
2mg
50mg
5mg
Product Code. Quantity unitSize
16487176 100 mg 100mg
16479717 5 mg 5mg
16477188 10 mg 10mg
16440286 2 mg 2mg
16403217 50 mg 50mg
5 options
This item is not returnable. View return policy

Product Code. 16487176

Brand: Medchem Express HY16774

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Vericiguat (BAY1021189) is a potent, orally available and soluble guanylate cyclase stimulator.

Vericiguat (BAY1021189) is a potent, orally available and soluble guanylate cyclase stimulator.

Specifications

Chemical Name or Material Vericiguat
CAS 1350653-20-1
Color Light Brown
Purity Grade Notes Research
Molecular Formula C19H16F2N8O2
Quantity 100 mg
Synonym BAY1021189
Solubility Information DMSO : 60 mg/mL (140.72 mM; Need ultrasonic)
SMILES O=C(OC)NC1=C(N)N=C(C2=NN(CC3=CC=CC=C3F)C4=NC=C(F)C=C42)N=C1N
Molecular Weight (g/mol) 426.38
Formula Weight 426.38
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.