accessibility menu, dialog, popup

UserName

CAS RN 1323966-41-1

IUPAC Name: 4-fluoro-2-(trifluoromethoxy)benzamide
Synonyms: 4-fluoro-2-(trifluoromethoxy)benzamide
Molecular Weight (g/mol): 223.13
Molecular Formula: C8H5F4NO2
InChi Key: MNJBQYPDEXYTJF-UHFFFAOYSA-N
SMILES: NC(=O)C1=C(OC(F)(F)F)C=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results