accessibility menu, dialog, popup

UserName

CAS RN 1323966-34-2

CH3OFNH2Cl

CAS RN 1323966-34-2

IUPAC Name: 1-(4-chloro-2-fluoro-3-methoxyphenyl)methanamine
Synonyms: 1-(4-chloro-2-fluoro-3-methoxyphenyl)methanamine
Molecular Weight (g/mol): 189.61
Molecular Formula: C8H9ClFNO
InChi Key: OIGDUJMZHJQGMX-UHFFFAOYSA-N
SMILES: COC1=C(Cl)C=CC(CN)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results