CAS RN 131570-56-4
IUPAC Name:
2-{[(tert-butoxy)carbonyl]amino}-3-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)propanoic acid
Synonyms:
2-[(tert-butoxycarbonyl)amino]-3-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}propanoic acid
Molecular Weight (g/mol):
426.47
Molecular Formula:
C23H26N2O6
InChi Key:
MVWPBNQGEGBGRF-UHFFFAOYNA-N
SMILES:
CC(C)(C)OC(=O)NC(CNC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
1
–
1
of
1
results
1
(R)-2-(Boc-amino)-3-(Fmoc-amino)propionic acid, 95%, Thermo Scientific™
CAS: 131570-56-4 Molecular Formula: C23H26N2O6 Molecular Weight (g/mol): 426.469 MDL Number: MFCD00236844 InChI Key: MVWPBNQGEGBGRF-LJQANCHMSA-N Synonym: boc-d-dap fmoc-oh,boc-n3-fmoc-d-2,3-diaminopropionic acid,r-3-9h-fluoren-9-yl methoxy carbonyl amino-2-tert-butoxycarbonyl amino propanoic acid,n-boc-n'-fmoc-d-2,3-diaminopropionic acid,boc-d-dpr fmoc-oh,ambotzbaa1031,boc-d-dapa fmoc-oh,pubchem14750,rarechem bk pt 0245,boc-d-dapa fmoc-oh.solvent PubChem CID: 2755948 IUPAC Name: (2R)-3-(9H-fluoren-9-ylmethoxycarbonylamino)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid SMILES: CC(C)(C)OC(=O)NC(CNC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13)C(=O)O