accessibility menu, dialog, popup

UserName

CAS RN 131185-00-7

CH3NNHSH2N

CAS RN 131185-00-7

IUPAC Name: (5-methylpyridin-2-yl)thiourea
Synonyms: 5-methylpyridin-2-ylthiourea
Molecular Weight (g/mol): 167.23
Molecular Formula: C7H9N3S
InChi Key: GEYNPFWKKUCREI-UHFFFAOYSA-N
SMILES: CC1=CN=C(NC(N)=S)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results