accessibility menu, dialog, popup

UserName

OHONH2SFFFFF

CAS RN 1266124-33-7

IUPAC Name: 2-amino-3-[4-(pentafluoro-λ⁶-sulfanyl)phenyl]propanoic acid
Synonyms: 2-amino-3-[4-(pentafluoro-λ⁶-sulfanyl)phenyl]propanoic acid
Molecular Weight (g/mol): 291.24
Molecular Formula: C9H10F5NO2S
InChi Key: AOHKRFDNEZJUEK-UHFFFAOYNA-N
SMILES: NC(CC1=CC=C(C=C1)S(F)(F)(F)(F)F)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results