accessibility menu, dialog, popup

UserName

CH3OFFFN

CAS RN 1261679-13-3

IUPAC Name: 2-methyl-5-(trifluoromethoxy)benzonitrile
Synonyms: 2-methyl-5-(trifluoromethoxy)benzonitrile
Molecular Weight (g/mol): 201.15
Molecular Formula: C9H6F3NO
InChi Key: VZVKYRYUNZHYII-UHFFFAOYSA-N
SMILES: CC1=C(C=C(OC(F)(F)F)C=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results