CAS RN 1253186-56-9
CAS RN 1253186-56-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
GSK-7975A, MedChemExpress
MedChemExpress GSK-7975A is a potent and orally available CRAC channel inhibitor.