Learn More
GSK-7975A, MedChemExpress
Product Code. 16471437
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
Product Code. 16471437
Brand: Medchem Express HY12507
MedChemExpress GSK-7975A is a potent and orally available CRAC channel inhibitor.
GSK-7975A is a potent and orally available CRAC channel inhibitor.
Specifications
Chemical Name or Material GSK-7975A
CAS 1253186-56-9
Color White
Purity Grade Notes Research
Molecular Formula C18H12F5N3O2
Quantity 50 mg
Solubility Information DMSO : ≥ 90 mg/mL (226.53 mM)
SMILES O=C(NC1=NN(CC2=CC=C(O)C=C2C(F)(F)F)C=C1)C3=C(F)C=CC=C3F
Molecular Weight (g/mol) 397.3
Formula Weight 397.3
Percent Purity 98.24%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.