CAS RN 1188910-76-0
CAS RN 1188910-76-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Agerafenib, MedChemExpress
MedChemExpress Agerafenib (CEP-32496; RXDX-105) is a highly potent and orally efficacious inhibitor of BRAFV600E with a Kd of 14 nM.