Learn More
Agerafenib, MedChemExpress
Product Code. 16476666
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16476666
Brand: Medchem Express HY15200
MedChemExpress Agerafenib (CEP-32496; RXDX-105) is a highly potent and orally efficacious inhibitor of BRAFV600E with a Kd of 14 nM.
Agerafenib (CEP-32496
Specifications
Chemical Name or Material Agerafenib
CAS 1188910-76-0
Color White
Purity Grade Notes Research
Molecular Formula C24H22F3N5O5
Quantity 100 mg
Synonym CEP-32496 RXDX-105
Solubility Information DMSO : 50 mg/mL (96.63 mM; Need ultrasonic)
SMILES O=C(NC1=NOC(C(C)(C)C(F)(F)F)=C1)NC2=CC=CC(OC3=C(C=C(OC)C(OC)=C4)C4=NC=N3)=C2
Molecular Weight (g/mol) 517.46
Formula Weight 517.46
Percent Purity 99.53%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.