accessibility menu, dialog, popup

UserName

CAS RN 1073354-81-0

BOOH3CCH3H3CH3CNF

CAS RN 1073354-81-0

IUPAC Name: 2-(4-fluorophenyl)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Synonyms: 2-(4-fluorophenyl)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Molecular Weight (g/mol): 299.15
Molecular Formula: C17H19BFNO2
InChi Key: CTCLIEHCPOJNSW-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CN=C(C=C1)C1=CC=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results