Assay Details
| Gene Symbol: LOC100144966 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126532.1 | NP_001120004.1 | 5 - 6 | 572 | 81 | |
| GenBank mRNA | BC158287.1 | - | 5 - 6 | 572 | 81 | |
More Information
Set Membership: |
|

LOC100144966
Genomic Map