Assay Details
| Gene Symbol: LOC4346747 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795965.2 | XP_015651451.1 | 8 - 9 | 893 | 83 | |
| GenBank mRNA | AK068681.1 | - | 8 - 9 | 848 | 83 | |
More Information
Set Membership: |
|

LOC4346747
Genomic Map