Assay Details
| Gene Symbol: LOC4347499 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795738.2 | XP_015651224.1 | 1 | 1683 | 151 | |
| GenBank mRNA | AK068607.1 | - | 1 | 1658 | 151 | |
More Information
Set Membership: |
|

LOC4347499
Genomic Map