Assay Details
| Gene Symbol: TOGARAM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012099164.3 | XP_011954554.3 | 5 - 6 | 3081 | 126 | |
| XM_027957299.1 | XP_027813100.1 | 5 - 6 | 3081 | 126 | ||
More Information
Set Membership: |
|

TOGARAM1
Genomic Map