Assay Details
| Gene Symbol: GRPR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004021935.4 | XP_004021984.1 | 2 - 3 | 766 | 76 | |
| GenBank mRNA | EU487010.1 | - | 1 - 2 | 271 | 76 | |
More Information
Set Membership: |
|

GRPR
Genomic Map