Assay Details
| Gene Symbol: A930028O11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK020901.1 | - | 1 | 1048 | 68 | |
| BB280937.1 | - | 1 | 238 | 68 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | seven in absentia 2 | Siah2 |
AA415433, Sinh2 |
-5.2kb | Anti-sense | Chr. 3 - 58674949 - 58692388 on NCBI Build GRCm38 |
| Downstream |
family with sequence similarity 188, member B2 |
Fam188b2 |
5330432B20Rik, BB026216 |
91.4kb | Anti-sense | Chr. 3 - 58790117 - 58816782 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

A930028O11Rik
Genomic Map