Learn More
Abnova™ ZYG11B Recombinant Protein
Product Code. p-3723561
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
Product Code. 16124373
Brand: Abnova™ H00079699P01.10ug
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Specifications
Accession Number AAH29832
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
Gene ID (Entrez) 79699
Molecular Weight (g/mol) 68.64
Name ZYG11B (Human) Recombinant Protein (P01)
pH Range 8
Preparation Method In vitro wheat germ expression system
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Source Wheat Germ (in vitro)
Immunogen MEKTKPEILKLVVTGMRNHPMNLPVQLAASACVFNLTKQDLAAGMPVRLLADVTHLLLKAMEHFPNHQQLQKNCLLSLCSDRILQDVPFNRFEAAKLVMQWLCNHEDQNMQRMAVAIISILAAKLSTEQTAQLGTELFIVRQLLQIVKQKTNQNSVDTTLKFTLSALWNLTDESPTTCRHFIENQGLELFMRVLESFPTESSIQQKVLGLLNNIAEVQELHSELMWKDFIDHISSLLHSVEVEVSYFAAGIIAHLISRGEQAWTLSRSQRNSLLDDLHSAILKWPTPECEMVAYRSFNPFFPLLGCFTTPGVQLWAVWAMQHVCSKNPSRYCSMLIEEGGLQHLYNIKDHEHTDPHVQQIAVAILDSLEKHIVRHGRPPPCKKQPQARLN
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Gene Alias FLJ13456/ZYG11
Common Name ZYG11B
Gene Symbol ZYG11B
Cross Reactivity Human
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Form Solution