Description
Essential component of the mitotic checkpoint, which prevents cells from prematurely exiting mitosis. Required for the assembly of the dynein-dynactin and MAD1-MAD2 complexes onto kinetochores.
Specifications
Specifications
| Antigen | ZWILCH |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.27 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 |
| Gene | ZWILCH |
| Gene Accession No. | Q8R060, Q9H900 |
| Gene Alias | hZwilch, KNTC1AP, Protein zwilch homolog, ZWILCH |
| Gene Symbols | ZWILCH |
| Show More |