Learn More
Invitrogen™ ZOOM™ IPGRunner™ Cassettes and Trays
Product Code. 12000216
Click to view available options
Quantity:
10 trays, 10 cassette(s)
Unit Size:
Pack of 10
Product Code. 12000216
Brand: Invitrogen™ ZM0008
Includes
Box of 10 cassettes and a pack of 10 trays
Convenient, disposable trays
ZOOM Equilibration Trays are designed for use with the ZOOM IPGRunner Cassettes to perform equilibration of ZOOM Strips after first-dimension isoelectric focusing and prior to second-dimension (2D) SDS-PAGE.
  • Simplify the equilibration and incubation process in 2D electrophoresis
  • Savings in time and labor
  • Optimal equilibration of samples
  • Minimal risk of sample contamination and damage to IPG strips
2D Gel Electrophoresis, IsoElectric Focusing, Protein Gel Electrophoresis, Protein Sample Fractionation, Protein Sample Preparation and Protein Purification, Proteins, Expression, Isolation and Analysis
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage The ZOOM™ Equilibration Trays are supplied in a pack of 10. The combo kit of ZOOM™Cassettes and ZOOM™ Equilibration Trays includes a box of 10 cassettes and a pack of 10 trays. Store at room temperature. Guaranteed stable for one year when properly stored.
For Use With (Equipment) ZOOM IPGRunner System
Product Line IPGRunner, ZOOM
Quantity 10 trays, 10 cassette(s)