Learn More
Buerkle™ Zone Sampler Spiralus
Product Code. 10532138 Shop All Bürkle Products
Change view
Click to view available options
Capacity (Metric):
770 mL
1100 mL
Length (Metric):
145 cm
200 cm
Unit Size:
Each
Product Code. Capacity (Metric) Length (Metric) unitSize
10532138 770 mL 145 cm Each
11857251 1100 mL 200 cm Each
2 options
Product Code. 10532138
Brand: Buerkle™ 53180145
Only null left
Add to basket
Add to basket
Buerkle™ Zone Sampler Spiralus provides an excellent all-layer sample.
  • Representative sample of the container layers.
  • aluminum, anodised
  • Tip PA
  • Low weight
TRUSTED_SUSTAINABILITY
Specifications
Product Type Zone Sampler Spiralus
Material Aluminum
Includes 8 openings
Capacity (English) 26 oz.
Capacity (Metric) 770 mL
For Use With (Application) Sampling Coarse-Grained Bulk Goods, Seeds and Pellets at Locations Such as Soil Heaps, Warehouses, Trucks, Railway Goods Wagons
Diameter (Metric) Outer 38 mm
Length (Metric) 145 cm