Learn More
Zomepirac sodium salt, MedChemExpress
Product Code. 16468486
Encompass_Preferred
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
Product Code. 16468486
Brand: Medchem Express HYB0890
MedChemExpress Zomepirac sodium salt (McN-2783-21-98) is a potent prostaglandin biosynthesis inhibitor. Zomepirac sodium salt is a non-steroidal anti-inflammatory drug (NSAID). Zomepirac sodium salt can cause immune-mediated liver injury.
Zomepirac sodium salt (McN-2783-21-98) is a potent prostaglandin biosynthesis inhibitor. Zomepirac sodium salt is a non-steroidal anti-inflammatory drug (NSAID). Zomepirac sodium salt can cause immune-mediated liver injury.
Specifications
Chemical Name or Material Zomepirac sodium salt
CAS 64092-48-4
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C15H13ClNNaO3
Quantity 100 mg
Synonym McN-2783-21-98
Solubility Information DMSO : 100 mg/mL (318.77 mM; Need ultrasonic) ∣H2O : 5 mg/mL (15.94 mM; Need ultrasonic)
SMILES O=C(O[Na])CC1=CC(C)=C(C(C2=CC=C(Cl)C=C2)=O)N1C
Molecular Weight (g/mol) 313.71
Formula Weight 313.71
Percent Purity 99.42%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.