Learn More
Zofenopril calcium, MedChemExpress
Product Code. 16411038
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. Quantity unitSize
16411038 5 mg 5mg
16458406 100 mg 100mg
16444437 50 mg 50mg
16428568 10 mg 10mg
4 options
Product Code. 16411038
Brand: Medchem Express HYB0655
MedChemExpress Zofenopril Calcium (SQ26991) is an antioxidant that acts as an angiotensin-converting enzyme inhibitor.
Zofenopril Calcium (SQ26991) is an antioxidant that acts as an angiotensin-converting enzyme inhibitor.
Specifications
Chemical Name or Material Zofenopril calcium
CAS 81938-43-4
Color White
Purity Grade Notes Research
Molecular Formula C22H22Ca0.5NO4S2
Quantity 5 mg
Synonym SQ26991
Solubility Information DMSO : 2 mg/mL (4.46 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C(N(C[C@@H](SC1=CC=CC=C1)C2)[C@@H]2C([O-])=O)[C@H](C)CSC(C3=CC=CC=C3)=O.[0.5Ca2+]
Molecular Weight (g/mol) 448.58
Formula Weight 448.58
Percent Purity 99.56%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration