Learn More
ZNHIT6 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 28019-1-AP-150UL
 View more versions of this product
Product Code. 16835955
  • 4836.10 NOK / 150µL
Description
The ZNHIT6 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human ZNHIT6. This antibody recognizes human, mouse antigen. The ZNHIT6 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
ZNHIT6
Polyclonal
Unconjugated
150 μL
54680
IgG
Western Blot, ELISA
500 μg/mL
Rabbit
Primary
Human, Mouse