Learn More
ZNHIT6 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 28019-1-AP-20UL
This item is not returnable.
View return policy
Description
The ZNHIT6 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human ZNHIT6. This antibody recognizes human, mouse antigen. The ZNHIT6 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| ZNHIT6 | |
| Polyclonal | |
| Unconjugated | |
| 20 μL | |
| 54680 | |
| IgG |
| Western Blot, ELISA | |
| 500 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse |