missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF774 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25449-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
May be involved in transcriptional regulation.Specifications
ZNF774 | |
Polyclonal | |
Unconjugated | |
ZNF774 | |
zinc finger protein 774, ZNF774 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
342132 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.42 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q6NX45 | |
ZNF774 | |
ZNF774 Fusion Protein Ag22300 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |