missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF570 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27811-1-AP-150UL
This item is not returnable.
View return policy
Description
ZNF570 is a new candidate transcription factor.Specifications
| ZNF570 | |
| Polyclonal | |
| Unconjugated | |
| ZNF570 | |
| zinc finger protein 570, ZNF570 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 148268 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.43 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q96NI8 | |
| ZNF570 | |
| ZNF570 Fusion Protein Ag26925 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction