missing translation for 'onlineSavingsMsg'
Learn More

ZNF570 Rabbit anti-Human, Polyclonal, Proteintech

Product Code. 16845845 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16845845 20 μL 20µL
16835845 150 μL 150µL
2 options
This item is not returnable. View return policy

Product Code. 16845845

Brand: Proteintech 278111AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ZNF570 is a new candidate transcription factor.

Specifications

Antigen ZNF570
Applications Western Blot
Classification Polyclonal
Concentration 0.43 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene ZNF570
Gene Accession No. Q96NI8
Gene Alias zinc finger protein 570, ZNF570
Gene Symbols ZNF570
Host Species Rabbit
Immunogen ZNF570 Fusion Protein Ag26925
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 148268
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.