Learn More
ZNF549 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 25592-1-AP-150UL
 View more versions of this product
Product Code. 16834155
  • 4836.10 NOK / 150µL
Description
ZNF549 may be involved in transcriptional regulation.
Specifications
Specifications
ZNF549
Polyclonal
Unconjugated
ZNF549
zinc finger protein 549, ZNF549
Rabbit
Antigen Affinity Chromatography
RUO
256051
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Western Blot
0.25 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q6P9A3
ZNF549
ZNF549 Fusion Protein Ag22371